missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Attune™ NxT Windows 10 OS Upgrade Kit

Product Code. 16586250
Change view
Click to view available options
Quantity:
1 kit
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16586250 1 kit Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16586250 Supplier Invitrogen™ Supplier No. A41324
 Restricted Item

This item requires a price update prior to being ordered.

Request a Price for this item here.

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

The Attune NxT Windows 10 OS Upgrade Kit is for use with the Attune NxT Acoustic Focusing Cytometer.

It allows the Attune NxT computer [model Optiplex XE2 or Optiplex XE2 (2x2TB)] to be reimaged to run the data collection and analysis software using the Microsoft Windows 10 operating system (OS).

TRUSTED_SUSTAINABILITY

Specifications

Product Line Attune
Content And Storage • 1 Windows 10 license
• 1 Windows 10 computer image

Quantity 1 kit
Shipping Condition Room Temperature
For Use With (Equipment) Attune Acoustic Focusing Cytometer
Product Type Software

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.