missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Attune™ Shutdown Solution (1X)

Product Code. 17334743
Change view
Click to view available options
Quantity:
250 mL
1 L
Unit Size:
1 L
250mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
17334743 1 L 1 L
15351086 250 mL 250mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 17334743 Supplier Invitrogen™ Supplier No. J106628
 Restricted Item
In Stock
Add to basket
Edge
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Attune™ Shutdown Solution (1X)

  • A cleaning buffer that is used in the automated shutdown procedure for the Attune™ Acoustic Focusing Cytometer
  • Azide-free shutdown solution has been formulated to minimize bubble formulation that can occur in the system lines overnight
  • Less bubbles help keep the fluid lines clean, and minimize microbial growth in the fluid lines
  • Use at the end of each day to ensure that all sample fluids and dyes have been removed from the system
  • Ensure that fluidic lines and pumps are filled with Attune™ Shutdown Solution to prevent salt deposits from forming in the lines
  • Shutdown takes approximately 20 minutes, but most of the steps are automatic and under the control of the Attune™ Cytometric Software
  • Shutdown procedures should be run at least once a day, even if the instrument is intended for continuous use
  • Proper cleaning of the instrument ensures its consistent and accurate operation
  • The ready-to-use 1X shutdown solution was designed to eliminate the need for dilution prior to use, as well as access to a purified water system
  • The Attune™ Shutdown Solution is prepared, filtered, and packaged in a sterile environment to ensure the highest quality
  • Each batch is traceable by its lot number designation and has been validated for use on the Attune™ Acoustic Focusing Cytometer
TRUSTED_SUSTAINABILITY

Specifications

Product Line Attune
Content And Storage Store at room temperature.
Quantity 1 L
For Use With (Equipment) Attune Acoustic Focusing Cytometer
Product Type Shutdown Solution

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.