Learn More
Invitrogen™ Attune™ Xenith™ Fluidics Umbilical Tubing
Product Code. 31759267
Change view
Click to view available options
Product Type:
Bleach Tubing Quick Connect
Ground Strap
Umbilical Bleach Line
Umbilical Electrical Cable
Umbilical Sheath Line
Umbilical Shut Down Line
Umbilical Sleeve
Umbilical Wash Line
Umbilical Waste Line
Unit Size:
Each
9 product options available for selection
Product selection table with 9 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Product Type Unit Size
31759267 Bleach Tubing Quick Connect Each
31759158 Ground Strap Each
31759123 Umbilical Electrical Cable Each
31759234 Umbilical Sleeve Each
31759265 Umbilical Waste Line Each
31759190 Umbilical Shut Down Line Each
31759177 Umbilical Wash Line Each
31759171 Umbilical Bleach Line Each
31759266 Umbilical Sheath Line Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 9 options available.
9 options
Product Code. 31759267 Supplier Invitrogen™ Supplier No. 100125295CS
Estimated Shipment: 20-11-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
These parts are replacements for components within the umbilical tubing that carries fluid between the fluid cart and the Attune Xenith Flow Cytometer.
The Invitrogen Attune Xenith Flow Cytometer is a spectrally enabled system that provides clog-resistant performance with acoustic-assisted hydrodynamic focusing. The system has an external fluid cart that holds all the fluids required for everyday use of the system. These parts are replacements of components within the umbilical tubing that carries fluidics between the cart and the instrument.
TRUSTED_SUSTAINABILITY
Specifications
Quantity 1 Ea.
For Use With (Equipment) Attune Xenith Flow Cytometer
Product Type Bleach Tubing Quick Connect