Learn More
Invitrogen™ Attune™ Xenith™ SIP Shield
Product Code. 31759045
Change view
Click to view available options
Quantity:
1 Ea.
Unit Size:
Each
Product Code. Quantity Unit Size
31759045 1 Ea. Each
1 options
Product Code. 31759045 Supplier Invitrogen™ Supplier No. 100101323CS
Estimated Shipment: 02-10-2026
Add to basket
Add to basket
The Attune Xenith SIP Shield is a key part of instrument safety and is available as a replacement should any damage occur to the existing part provided with the instrument.
The Attune Xenith SIP Shield is for use with the Attune Xenith Flow Cytometer and is a key part of instrument safety, providing a barrier between the user and the sample during acquisition of data. This replacement part is available should any damage occur to the existing part provided with the instrument.
TRUSTED_SUSTAINABILITY
Specifications
Quantity 1 Ea.
For Use With (Equipment) Attune Xenith Flow Cytometer
Product Type SIP Shield