Learn More
3M™ Aura™ 9322+ Particulate Respirators
Provides protection against fine dusts and water-based mists.
Brand:  3M™ 7100004175
Product Code. 11747574
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »
Description
Description
  • Unique three-panel design for greater comfort and easy communication
  • Soft cover web on inner panel ensures greater comfort
  • Centrally positioned cool flow valve reduces heat build-up and makes breathing easier and cooler
  • Evenly tensioned headbands to relieve strap pressure
  • Lightweight and foldable
  • Individually packed to avoid contamination during storage
  • FFP2 level protection
  • Particulates Assigned Protection Factor of 10 (NPF 12)