missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Pro-Lab Diagnostics™ Auramine Phenol Stain
Click to view available options
Quantity:
500 mL
Specifications
Specifications
| Type | Auramine Stain |
| Quantity | 500 mL |
| Certifications/Compliance | CE marked (98/78/EC) |
| Format | Liquid |
| Color | Yellow |
| Composition | Phenol 2.5 - <5% , Ethanol 2.5 - <5% , 4,4'-Carbonimidoylbis(N,N-dimethylaniline) monohydrochloride 0.25 - <0.5% , Methanol 0.25 - <0.5% |
| Storage Requirements | Keep at temperature not exceeding 20°C |
| Stain Type | Acid Fast |
| Volume (Metric) | 500 mL |
Safety and Handling
Product Identifier
- Auramine Phenol
- Danger
- Skin corrosion/irritation Category 1 B
- Serious eye damage/eye irritation Category 1
- Germ cell mutagenicity Category 2
- LONG-TERM AQUATIC HAZARD Chronic 3
- H314-Causes severe skin burns and eye damage.
- H341-Suspected of causing genetic defects.
- H412-Harmful to aquatic life with long lasting effects.
- P273-Avoid release to the environment.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
- P501-Dispose of contents/container to .
- P202-Do not handle until all safety precautions have been read and understood.
- P260-Do not breathe dust/fume/gas/mist/vapours/spray.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P363-Wash contaminated clothing before reuse.
- P405-Store locked up.
- MIXTURE LIST-Contains : phenol, ethanol, 4,4'-carbonimidoylbis(N, N-dimethylaniline)monohydrochloride,methanol
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction