missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
AURKC (Aurora C) is a chromosomal passenger protein that forms complexes with AURKB (Aurora B) and inner centromere proteins, playing a role in organizing microtubules in relation to centrosome⁄spindle function during mitosis. It is overexpressed in several cancer cell lines, suggesting an involvement in oncogenic signal transduction.Specifications
AAH75064 | |
Protein assays and analysis | |
Frozen | |
AURKC (Aurora C) Recombinant Human Protein | |
1mg | |
For research use only. Not for use in diagnostic procedures | |
AURKC (Aurora C), Aurora A, STK13 | |
AURKC (aurora C) was pre-diluted in enzyme dilution buffer and assayed in 25mM Tris (pH 7.5), 10mM MgCl2, 0.5mM EGTA, 0.5mM Na3VO4, 5mM β-glycerophosphate, 2.5mM DTT, 0.01% Triton™ X-100, 200μM ATP, 200μg/mL kemptide peptide substrate (LRRASLG) and trace [ 32P]-γ-ATP for 10 minutes at 30°C | |
AURKC (Aurora C) Recombinant Human Protein | |
Human | |
Baculovirus |
0.45mg/mL | |
Frozen | |
61.7kDa | |
90% by SDS-PAGE | |
–80°C | |
1 Each | |
AURKC (Aurora C) Recombinant Human Protein | |
Aurora | |
37nmoles of phosphate transferred to Kemptide peptide substrate (LRRASLG) per minute per mg of total protein at 30°C. Activity determined at a final protein concentration of 8.3μg/mL | |
GST Tag | |
Kinase assay |
For Research Use Only. Not for use in diagnostic procedures.