Learn More
Pacific Handy Cutter Auto-Retractable Metal Safety Knife
Product Code. 17769027
Change view
Click to view available options
Blade Material:
Aluminum
Unit Size:
Pack of 10
Product Code. Blade Material Unit Size
17769027 Aluminum Pack of 10
1 options
Product Code. 17769027 Supplier Pacific Handy Cutter Supplier No. QBAR
  • ONLINE EXCLUSIVE PRICE
    328.00 €
    300.77 € / Pack of 10
    Save 27.23 €
    8% Off
Estimated Shipment: 30-10-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
The QBAR is a particularly robust safety knife made of aluminum for the toughest applications! It has a fully automatic blade retraction and is therefore particularly safe.
  • In addition, it is equipped as standard with a blade with a rounded tip to reduce the risk of puncturing.
  • The blade retracts fully automatically into the body, even if your thumb remains on the slider after the cut.
  • Thanks to the three-sided ambidextrous slider, it is particularly ergonomic and can be used flexibly for horizontal and vertical cuts.
  • The sturdy metal handle has non-slip rubber grips.
  • Your knife is dirty? No problem. Thanks to the central lock, it can be opened and cleaned very easily without tools.
  • Change the blade in the same way. V
  • ery practically - you can accommodate replacement blades inside the handle.
  • The QBAR can be attached or hung by means of an eyelet at the end of the handle.
  • And the tape splitter on the underside of the knife head minimises use of your blade.
  • Especially suitable for thicker cutting materials, corrugated cardboard, strapping, adhesive tape, foil, textiles, felt, PVC, leather, rubber, string, etc.
  • QBAR is a premium quality safe cutting tool.
Specifications
Blade Material Aluminum
For Use With (Application) Cutting Thicker Cutting Materials, Corrugated Cardboard, Strapping, Adhesive Tape, Foil, Textiles, Felt, PVC, Leather, Rubber, String, Etc.
Disposable Reusable
Sterility Non-sterile