missing translation for 'onlineSavingsMsg'
Learn More

Bel-Art™ Biohazard Disposal Bags with Sterilization Indicator

Product Code. 10574434 Shop All SP Scienceware Products
Change view
Click to view available options
Capacity (Metric):
189 L
Unit Size:
Pack of 100
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Capacity (Metric) unitSize
10574434 189 L Pack of 100
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10574434 Supplier Bel-Art™ Supplier No. 0649408

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Resists tears and leaks. Bel-Art Products™ Biohazard Disposal Bagss with Sterilization Indicator are constructed of heavy-duty, high-impact polypropylene.

  • Red bags display the standard biohazard warning symbol and precautionary procedures in four languages
  • Thickness: 2mm
  • Blended polymer construction
  • Autoclavable to 121° C (250° F) with sterilization indicator patch that darkens when autoclaved

Specifications

Autoclavable Autoclavable
Certifications/Compliance ASTM 1709 Dart Impact, ASTM 1992 Tear Resistance
Color Red
Disposable Single-use
For Use With (Application) Biohazard waste
Includes Sterilization indicator, Biohazard warning labels in four languages
Material HDPE
Height (Metric) 122 mm
Capacity (Metric) 189 L
Width (Metric) 94 mm
Thickness (Metric) 0.500 mm
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.