missing translation for 'onlineSavingsMsg'
Learn More

ratiolab™ Autoclavable Polypropylene Bags

Product Code. 13511030
Change view
Click to view available options
Dimensions (L x W):
300 x 500 mm
800 x 600 mm
1100 x 700 mm
Capacity (Metric):
6 L
32 L
72 L
Unit Size:
Pack of 350
Pack of 500
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Dimensions (L x W) Capacity (Metric) Unit Size
13511030 300 x 500 mm 6 L Pack of 500
15518713 800 x 600 mm 32 L Pack of 500
15526939 1100 x 700 mm 72 L Pack of 350
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 13511030 Supplier ratiolab™ Supplier No. 7002100
Reliable Agri Testing Solutions
Use promo code 28024
View all promotions
In Stock
Add to basket
Add to basket
This item is not returnable. View return policy

This autoclavable, disposable waste bag is for the sterilization and disposal of hazardous and contaminated labware.

  • With BIOHAZARD warning Instruction and sterilization indicator patch
  • Safety imprint in German, English, French and Spanish languages
  • Rectangle white indicator field turns into dark when autoclaved
  • For autoclaving, the bag must not be completely closed
  • Autoclavable at 134°C for 20 min.
  • Tolerance: ±5%

Specifications

Autoclavable Autoclavable
Disposable Single-use
For Use With (Application) Waste Disposal
Includes Bag with Indicator Patch
Material Polypropylene
Quantity 500 Bags/Pk.
Thickness (English) 2 mil
Dimensions (L x W) 300 x 500 mm
Length (English) 19.7 in.
Width (English) 11.8 in.
Product Type Biohazard Disposal Bag
Capacity (English) 1.6 gal.
Capacity (Metric) 6 L
Length (Metric) 500 mm
Width (Metric) 300 mm
Thickness (Metric) 50 μm
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.