missing translation for 'onlineSavingsMsg'
Learn More

Masterflex™ Polypropylene Straight Connector Barbed Fitting

Product Code. 12418700 Shop All Masterflex Products
Change view
Click to view available options
For Use With:
1/16 in. tubing I.D.
1/8 in. tubing I.D.
Unit Size:
Pack of 25
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. For Use With Unit Size
12418700 1/16 in. tubing I.D. Pack of 25
12459570 1/8 in. tubing I.D. Pack of 25
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 12418700 Supplier Masterflex™ Supplier No. 0636511
  • ONLINE EXCLUSIVE PRICE
    38.85 €
    35.47 € / Pack of 25
    Save 3.38 €
    8% Off
Estimated Shipment: 29-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

These economical straight barbed fittings easily connect fluid paths in the laboratory without the use of tools.

  • Ideal for low-pressure applications, typically up to 30 psi (2 bar)
  • Chemical resistance
  • Slip flexible tubing or hose onto the barb to create a secure seal
  • No leaks, flow restrictions or costly downtime
  • Sterilization through atuoclaving or ethylene oxide (ETO)

Specifications

Autoclavable Autoclavable
Material Polypropylene
For Use With 1/16 in. tubing I.D.
Packaging Quantity 25/Pk.
Port Size 1/32 in. orifice; 25/32 x 1/4 in.
Sterilization Method Autoclave or Ethylene Oxide
Shape Straight
Includes Straight connector
Color Clear
Maximum Pressure 30 psi (2 bar)
Product Type Barbed Fitting
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.