Learn More
Justrite Automatic Extinguisher Panel Control
Product Code. 17722398
Change view
Click to view available options
Product Type:
Control Panel
Unit Size:
Each
Product Code. Product Type Unit Size
17722398 Control Panel Each
1 options
Product Code. 17722398 Supplier Justrite Supplier No. JCBSFPC000
  • ONLINE EXCLUSIVE PRICE
    3213.00 €
    2911.71 € / Pack of 1
    Save 301.29 €
    9% Off
Estimated Shipment: 30-09-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
Automatic extinguisher panel control with smoke sensor flashing light panel
  • Automatic extinguisher panel control with smoke sensor flashing light panel for Lithium Battery EN Safety Cabinets
Specifications
Includes Smoke Sensor
Weight (Metric) 5 kg
Product Type Control Panel