missing translation for 'onlineSavingsMsg'
Learn More

BRAND™ Dispenser, tilt measure without bottle

Product Code. 10788764 Shop All Brand Products
Change view
Click to view available options
Capacity (Metric):
5 mL
10 mL
20 mL
25 mL
50 mL
100 mL
Unit Size:
Each
Product Code. Capacity (Metric) unitSize
10788764 10 mL Each
10367582 5 mL Each
10787444 20 mL Each
10205712 25 mL Each
10582073 50 mL Each
10012392 100 mL Each
6 options
This item is not returnable. View return policy

Product Code. 10788764

Brand: BRAND™ 43058

Only null left
Add to basket
Add to basket
This item is not returnable. View return policy

Tilt dispenser is best suited for dispensing suspensions.

  • Dispenser head
  • Calibrated to deliver (TD, Ex)
  • Inscriptions in green enamel
  • Particularly well-suited for dispensing suspensions
TRUSTED_SUSTAINABILITY

Specifications

Material Borosilicate Glass 3.3
Description Without bottle
Capacity (Metric) 10 mL
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.