Learn More
BRAND™ Dispenser, tilt measure without bottle
Tilt dispenser is best suited for dispensing suspensions.
73.65 € - 122.00 €
Product Code | Brand | Capacity (Metric) | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Capacity (Metric) | Price | Quantity & Availability | |||||
10367582
|
BRAND™
43055 |
5 mL |
73.65 €
Each |
Estimated Shipment: 15-05-2024
Log in to see stock. |
|||||
10788764
|
BRAND™
43058 |
10 mL |
76.35 €
Each |
Estimated Shipment: 10-05-2024
Log in to see stock. |
|||||
10787444
|
BRAND™
43066 |
20 mL |
82.00 €
Each |
Estimated Shipment: 26-06-2024
Log in to see stock. |
|||||
10205712
|
BRAND™
43070 |
25 mL |
87.85 €
Each |
Estimated Shipment: 10-06-2024
Log in to see stock. |
|||||
10582073
|
BRAND™
43078 |
50 mL |
118.00 €
Each |
Estimated Shipment: 26-06-2024
Log in to see stock. |
|||||
10012392
|
BRAND™
43088 |
100 mL |
122.00 €
Each |
Estimated Shipment: 10-06-2024
Log in to see stock. |
|||||
Description
- Dispenser head
- Calibrated to deliver (TD, Ex)
- Inscriptions in green enamel
- Particularly well-suited for dispensing suspensions
Specifications
Without bottle |