missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Automation Tips for Beckman™ Liquid Handling Systems with MP20 and MP200 Dispense Heads

Product Code. 11781263 Shop All MBP Molecular BioProducts Products
Change view
Click to view available options
Volume (Metric):
20 μL
130 μL
200 μL
Filtered:
Filtered
Non-filtered
Sterility:
Non-sterile
Sterile
Fit for Purpose:
Certified Quality
Sterile
Sterile and PCR Ready
Unit Size:
Pack of 4800
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Volume (Metric) Filtered Sterility Fit for Purpose Unit Size
11781263 20 μL Filtered Sterile Sterile and PCR Ready Pack of 4800
11771263 20 μL Non-filtered Sterile Sterile Pack of 4800
11781223 20 μL Non-filtered Non-sterile Certified Quality Pack of 4800
11741253 130 μL Filtered Sterile Sterile and PCR Ready Pack of 4800
11751253 200 μL Non-filtered Sterile Sterile Pack of 4800
11791223 200 μL Non-filtered Non-sterile Certified Quality Pack of 4800
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
This item is not returnable. View return policy
Product Code. 11781263 Supplier Thermo Scientific™ Supplier No. 915021
Estimated Shipment: 03-03-2027
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Increase pipetting performance with Thermo Scientific™ Automation Tips for Beckman™ Liquid Handling Systems, which undergo a rigorous 15-point quality control program to ensure consistency.

Increase pipetting performance with Thermo Scientific™ Sterile Pipette Tips for Beckman™ Liquid Handling Systems, which undergo a rigorous 15-point quality control program to ensure consistency. These automation tips provide accurate and precise performance due to superior tip straightness and a low coefficient of variation. The pipette tips are validated on the actual workstations for which they are created.

Features:
  • Available with Thermo Scientific™ ART™ filter technology, low retention and extended length
  • Lot-tested during production on actual workstation for which they are created
  • Extended length tips compatible with 96-, 384-, and 1536-deep well pates
  • Low percentage coefficient of variation (%CV)
TRUSTED_SUSTAINABILITY

Specifications

Volume (Metric) 20 μL
Filtered Filtered
Sterility Sterile
Color Clear
Format Rack
Tip Style Extended Length
For Use With (Equipment) Beckman Coulter Liquid Handling Systems
Product Line Beckman
Fit for Purpose Sterile and PCR Ready
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.