missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Automation Tips for Caliper™ Zymark™ Liquid Handling Workstations
GREENER_CHOICE

Product Code. 11860421 Shop All MBP Molecular BioProducts Products
Click to view available options
Volume (Metric):
50 μL
80 μL
100 μL
150 μL
200 μL
Filtered:
Filtered
Non-filtered
Sterility:
Non-sterile
Sterile
Fit for Purpose:
Certified Quality
Sterile
Sterile and PCR Ready
Unit Size:
Pack of 4800
This item is not returnable. View return policy

Product Code. 11860421

Brand: Thermo Scientific™ 921262

Only null left
Add to basket
Add to basket
This item is not returnable. View return policy

Increase pipetting performance with these automation tips, which undergo a rigorous 15-point quality control program to ensure accuracy and consistency.

Increase pipetting performance with Thermo Scientific™ Automation Tips for Caliper™ Zymark™ Liquid Handling Workstations, which undergo a rigorous 15-point quality control program to ensure consistency. Their superior tip straightness and a low coefficient of variation provide accurate and precise performance. Compatible with: Caliper/Zymark RapidPlate, Caliper Presto, Allegro, Zephyr and SciClone.

Features:

TRUSTED_SUSTAINABILITY

Specifications

Volume (Metric) 100 μL
Filtered Non-filtered
Sterility Non-sterile
Color Clear
Format Rack
Tip Style Automation
For Use With (Equipment) Zymark/Caliper Liquid Handling Workstations
Product Line Caliper Zymark
Fit for Purpose Certified Quality
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.