Learn More
Thermo Scientific™ Automation Tips for PerkinElmer™/Packard™ Robotic Liquid Handling Systems with 96- and 384-Channel MDT Dispense Heads
GREENER_CHOICE
Product Code. 13508310 Shop All MBP Molecular BioProducts Products
Click to view available options
Volume (Metric):
10 μL
20 μL
30 μL
40 μL
50 μL
180 μL
235 μL
Autoclavable:
Autoclavable
Not Autoclavable
Filtered:
Filtered
Non-filtered
Fit for Purpose:
Certified Quality
Sterile
Sterile and PCR Ready
Unit Size:
Pack of 19200
Pack of 4800
Product Code. 13508310
Brand: Thermo Scientific™ 931261
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Increase pipetting effectiveness with these automation tips, achieving accurate and precise performance due to superior tip straightness and a low coefficient of variation.
Thermo Scientific™ Automation Tips for Perkin Elmer™/Packard™ Robotic Liquid Handling Systems with Varispan™ Dispense Arms, which undergo a rigorous 15-point quality control program to ensure consistency. This leads to accurate and precise performance due to superior tip straightness and a low coefficient of variation. The pipette tips are validated on the actual workstations for which they are created and the sterile pipette tips are certified free of RNase, DNase, DNA, endotoxin, bioburden and pyrogen.
  • Available in non-sterile, sterile, with Thermo Scientific™ ART™ filter technology and in low retention
  • Increased precision for the toughest multi-dispensing assays
  • Low percentage coefficient of variation (%CV)
TRUSTED_SUSTAINABILITY
Specifications
Volume (Metric) 30 μL
Filtered Non-filtered
Sterility Sterile
Color Clear
Autoclavable Not Autoclavable
Certifications/Compliance ISO 9001 and ISO 13485
Clean Claims Free of RNase, DNase, DNA, ATP, and endotoxin
Format Rack
Tip Style Automation
Packaging 96 tips/rack, 10 racks/pack, 5 packs/case
Quantity 4800 tips
For Use With (Equipment) Packard Liquid Handling System, PerkinElmer, Molecular Devices FLIPR Type A CCS 384-well Head, Molecular Devices FLIPR Type B Liberty 384-well Head
Product Line PerkinElmer/Packard
Fit for Purpose Sterile