missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Automation Tips for Tecan™ Liquid Handling Platforms
GREENER_CHOICE

Product Code. 10702953
Click to view available options
Volume (Metric):
10 μL
20 μL
50 μL
175 μL
200 μL
1000 μL
Filtered:
Filtered
Non-filtered
For Use With (Equipment):
Cavro, MiniPrep
Cavro, MiniPrep, PerkinElmer MultiProbe II HT, PerkinElmer MultiProbe II HT EX, PerkinElmer Janus
Cavro, Tecan Genesis, PerkinElmer MultiProbe II HT, PerkinElmer MultiProbe II HT EX, PerkinElmer Janus
Cavro, Tecan Genesis, Tecan Freedom EVO, PerkinElmer MultiProbe II HT EX, PerkinElmer Janus
Tecan Aquarius, Tecan TeMo, PerkinElmer MultiProbe II HT, PerkinElmer MultiProbe II HT EX, PerkinElmer Janus
Tecan Freedom EVO LiHa Head, Tecan Genesis
Tecan Freedom EVO LiHa Head, Tecan Genesis LiHa Head
Tecan Freedom EVO, MiniPrep, Cavro
Tecan Genesis, Tecan Freedom EVO, MiniPrep, Cavro
Tecan Genesis, Tecan Freedom EVO, MiniPrep, Cavro, PerkinElmer MultiProbe II HT, PerkinElmer MultiProbe II HT EX, PerkinElmer Janus
Fit for Purpose:
Certified Quality
PCR Ready
Sterile
Sterile and PCR Ready
missing translation for 'unitSize'
Pack of 4800
Pack of 960
This item is not returnable. View return policy

Product Code. 10702953

missing translation for 'mfr': Thermo Scientific™ 904011G

Only null left
Add to basket
Add to basket
This item is not returnable. View return policy

Benefit from Thermo Scientific™ Sterile Automation Tips for Tecan™ Liquid Handling Platforms. They increase pipetting performance through the superior tip straightness and low %CV of these pipette tips.

Get accuracy and precision with Thermo Scientific™ Sterile Automation Tips for Tecan™ Liquid Handling Platforms, which undergo a rigorous 15-point quality control program to ensure consistency. Their superior tip straightness and a low coefficient of variation provide accurate and precise performance.

  • Available as standard, filtered and/or liquid sensing
  • Certified free of RNase, DNase, DNA, endotoxin, bioburden and pyrogen
  • Lot-tested during production on actual workstation for which they are created
  • Fully conductive, providing increased pipetting performance
  • Low percentage coefficient of variation (%CV)
TRUSTED_SUSTAINABILITY

Specifications

Volume (Metric) 1000 μL
Filtered Filtered
Sterility Sterile
Color Black
Autoclavable Not Autoclavable
Certifications/Compliance ISO 9001 and ISO 13485
Clean Claims Free of RNase, DNase, DNA, ATP, and endotoxin
Format Rack
Tip Style Conductive
Packaging Reload Insert, 96 tips/insert, 10 inserts/pack
Quantity 960 Tips
For Use With (Equipment) Tecan Genesis, Tecan Freedom EVO, MiniPrep, Cavro
Product Line Tecan
Fit for Purpose Sterile and PCR Ready
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.