missing translation for 'onlineSavingsMsg'
Learn More

Axygen™ Automation Tips for Tecan™, Zymark™, Biotek™ and Dynamic Devices
BIOPHARMA

Product Code. 12752836 Shop All Axygen Scientific Products
Change view
Click to view available options
Sterility:
Non-sterile
Sterile
Volume (Metric):
50 μL
100 μL
180 μL
200 μL
205 μL
Filtered:
Filtered
Non-filtered
Unit Size:
Pack of 4800
Pack of 960
8 product options available for selection
Product selection table with 8 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Sterility Volume (Metric) Filtered unitSize
12752836 Sterile 200 μL Non-filtered Pack of 4800
10059499 Non-sterile 50 μL Non-filtered Pack of 4800
12736689 Sterile 180 μL Filtered Pack of 4800
12746689 Sterile 205 μL Filtered Pack of 4800
12756689 Sterile 50 μL Filtered Pack of 4800
11366284 Sterile 100 μL Filtered Pack of 960
10535950 Non-sterile 200 μL Non-filtered Pack of 4800
10810501 Non-sterile 100 μL Non-filtered Pack of 4800
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 8 options available.
8 options
This item is not returnable. View return policy
Product Code. 12752836 Supplier Axygen™ Supplier No. EV200RS
Only null left
Add to basket
Add to basket
This item is not returnable. View return policy

Ensure consistent performance and quality. Axygen™ Automation Tips for Tecan™, Zymark™, Biotek™ and Dynamic Devices are constructed of clear polypropylene in 96-well racks. Available in nonsterile/sterile, non-filtered/filtered, wide-bore and bottom-labeled options with volumes ranging from 50 to 200μL.

  • Comprehensive line of pipette tips for automated liquid handling systems
  • Manufactured to stringent specifications under strict process controls
  • Assembled via automation to ensure consistent performance and quality
  • Tips are certified RNase-/DNase-free and nonpyrogenic
  • Available in a range of volumes: 50, 100, 180 and 200μL
  • Compatible with the following:
    • Tecan™ Freedom Evo™, TeMo and Aquarius with MCA96 head
    • Zymark™/Caliper RapidPlate™, SciClone™ and Zephyr™ with HV96 head
    • Biotek™ Precision™ and Precision XS 8 or 12 head
    • Dynamic Devices Oasis LM 8/96-Channel-Standard Vol and 96-Channel-Low Vol head
TRUSTED_SUSTAINABILITY

Specifications

Volume (Metric) 200 μL
Filtered Non-filtered
Sterility Sterile
Color Clear
Rack Color Light Gray
Certifications/Compliance ANSI
Clean Claims DNase, Rnase Free
For Use With (Application) Non-pyrogenic, used for stringent specifications under strict process controls
Format Rack
Tip Style Automation, Round
Packaging SBS Rack
Quantity 50 Racks/Cs.
For Use With (Equipment) Tecan Freedom EVO, Tecan TeMo, Tecan Aquarius, Zymark/Caliper RapidPlate, SciClone, Zephyr, BioTek Precision, BioTek Precision XS, Dynamic Devices Oasis LM
Material Polypropylene
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.