Learn More
3M™ Auxiliary Air Supply Hose C-231
Product Code. 15749827
Change view
Click to view available options
Length (Metric):
1.2 m
Unit Size:
Pack of 6
Product Code. Length (Metric) unitSize
15749827 1.2 m Pack of 6
1 options
Product Code. 15749827 Supplier 3M™ Supplier No. 7000088331
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
3M™ Auxiliary Air Supply Hose C-231 provides a reliable connection for attaching Auxiliary air tools (e.g. spray guns) to the V-500 Regulator removing the need for multiple air hoses.
  • Removes the need for multiple air hoses
  • The hose is 1.2 m long
  • Fitted with 1/4 in. BSP thread fittings
TRUSTED_SUSTAINABILITY
Specifications
Connection Type 1/4 in. BSP Thread Fittings
Length (Metric) 1.2 m