missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Auxin

Product Code. 18796296
Change view
Click to view available options
Quantity:
50 mg
Unit Size:
50mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
18796296 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18796296 Supplier R&D Systems™ Supplier No. 6834/50
  • ONLINE EXCLUSIVE PRICE
    93.10 €
    84.00 € / 50mg
    Save 9.10 €
    9% Off
 Restricted Item
Estimated Shipment: 18-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

A Tocris product, part of R&D Systems. Chemical dimerizer used in auxin-inducible degron (AID) system; phytohormone

Auxin is a chemical dimerizer used in auxin-inducible degron (AID) systems. Induces degradation of a target protein tagged with the auxin-receptor F-box protein Tir1 E3 ligase AID in human colorectral cancer and mouse ES cells or tagged with AFB2 in A431 cells. Endogenous plant hormone.
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 1 H-Indole-3-acetic acid
CAS 87-51-4
Quantity 50 mg
Product Line Tocris
Target Protein Dimerizers
Molecular Formula C10H9NO2
Formula Weight 175.18
Purity 98%
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.