missing translation for 'onlineSavingsMsg'
Learn More

Auxinole, TRC

Product Code. 30134335
Change view
Click to view available options
Quantity:
50 mg
250 mg
500 mg
Unit Size:
250mg
500mg
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30134335 500 mg 500mg
30010986 50 mg 50mg
30012550 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30134335 Supplier Toronto Research Chemicals Supplier No. TRCA794595500MG
 Restricted Item
Estimated Shipment: 17-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 86445-22-9
Molecular Formula C20H19NO3
Molecular Weight (g/mol) 321.37
Synonym alpha-[2-(2,4-Dimethylphenyl)-2-oxoethyl]-1H-Indole-3-acetic Acid;
IUPAC Name 4-(2,4-dimethylphenyl)-2-(1H-indol-3-yl)-4-oxobutanoic acid
SMILES Cc1ccc(C(=O)CC(C(=O)O)c2c[nH]c3ccccc23)c(C)c1

Specifications

Quantity 500 mg
InChI Formula InChI=1S/C20H19NO3/c1-12-7-8-14(13(2)9-12)19(22)10-16(20(23)24)17-11-21-18-6-4-3-5-15(17)18/h3-9,11,16,21H,10H2,1-2H3,(H,23,24)
Formula Weight 321.1365
Percent Purity >95%
Chemical Name or Material Auxinole
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.