missing translation for 'onlineSavingsMsg'
Learn More

Avermectin B1, MedChemExpress

Product Code. 16476726
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16476726 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16476726 Supplier Medchem Express Supplier No. HY15311
 Restricted Item
Estimated Shipment: 08-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress Avermectin B1 (Abamectin) is a mixture of two similar segments of avermectin. Avermectin B1 is an orally anti-infection agent, which can be used in the research of parasitic worms, insect pests, agriculture and animal husbandry. Avermectin B1 can also induce the production of ROS and induces cytotoxicity, apoptosis and autophagy.

Avermectin B1 (Abamectin) is a mixture of two similar segments of avermectin. Avermectin B1 is an orally anti-infection agent, which can be used in the research of parasitic worms, insect pests, agriculture and animal husbandry. Avermectin B1 can also induce the production of ROS and induces cytotoxicity, apoptosis and autophagy.

Specifications

Chemical Name or Material Avermectin B1
CAS 71751-41-2
Color White
Purity Grade Notes Research
Molecular Formula C95H142O28
Quantity 100 mg
Synonym Abamectin Avermectin B1a-Avermectin B1b mixt.
Solubility Information DMSO : ≥ 247 mg/mL (142.60 mM)
SMILES C[C@@H](CC)[C@]1([H])[C@@H](C)C=C[C@@]2(O[C@@]3([H])C[C@]([H])(OC([C@@]4([H])[C@@](/C(CO5)=C/C=C/[C@H](C)[C@H](O[C@@]6([H])C[C@H](OC)[C@@H](O[C@]7([H])O[C@@H](C)[C@H](O)[C@@H](OC)C7)[C@H](C)O6)/C(C)=C/C3)(O)[C@@]5([H])[C@H](O)C(C)=C4)=O)C2)O1.C[C@H]8C=C[C@@]9(O[C@@]%10([H])C[C@]([H])(OC([C@@]%11([H])[C@@](/C(CO%12)=C/C=C/[C@H](C)[C@H](O[C@@]%13([H])C[C@H](OC)[C@@H](O[C@]%14([H])O[C@@H](C)[C@H](O)[C@@H](OC)C%14)[C@H](C)O%13)/C(C)=C/C%10)(O)[C@@]%12([H])[C@H](O)C(C)=C%11)=O)C9)O[C@@H]8C(C)C
Molecular Weight (g/mol) 1732.13
Formula Weight 1732.13
Percent Purity 96.89%
Grade Research
Physical Form Solid
For Use With (Application) COVID-19-immunoregulation
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.