missing translation for 'onlineSavingsMsg'
Learn More

Applied Biosystems™ Axiom™ SwiftArray™ Reagent Kit

Product Code. 31782930 Shop All Applied Biosystems Products
Change view
Click to view available options
No. of Samples:
1x96F
2x96F
Unit Size:
Each
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. No. of Samples Unit Size
31782930 2x96F Each
31782297 1x96F Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 31782930 Supplier Applied Biosystems™ Supplier No. 952636
 Restricted Item
Estimated Shipment: 18-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

The Axiom SwiftArray Reagent Kit is an essential processing component designed specifically for use with SwiftArrayStudio custom arrays. Available in two configurations—with wash or without—this reagent kit enables the processing of SwiftArrayStudio custom arrays while delivering industry-leading performance metrics.

The Axiom SwiftArray Reagent Kit is an essential processing component designed specifically for use with SwiftArrayStudio custom arrays. Available in two configurations—with wash or without—this reagent kit enables the processing of SwiftArrayStudio custom arrays while delivering industry-leading performance metrics.

The kit distinguishes itself through superior turnaround time, achieving a 60% improvement compared to the Axiom 2.0, and 33% improvement to the Illumina workflow, while simultaneously reducing required touchpoints and providing a more streamlined, user-friendly workflow. The reagent system incorporates the most current array content organized by application, offering end users enhanced flexibility to accommodate diverse experimental requirements.

The SwiftArray Reagent Kit functions exclusively in conjunction with SwiftArrayStudio Arrays, with typical applications involving custom array configurations. This mandatory pairing ensures optimal performance and compatibility within the SwiftArrayStudio ecosystem.

Features:

  • Rapid Processing Capability: Delivers a 2-day turnaround time from initiation to completion, enabling accelerated research workflows and time-sensitive project requirements
  • Enhanced Robustness: Engineered for reliable performance under demanding operational conditions, ensuring consistent results across varied experimental parameters and environmental factors
  • Adaptive Flexibility: Designed with modular architecture to accommodate diverse research applications and methodological approaches, allowing for customization based on specific scientific requirements
  • Sustainable Design: Incorporates environmentally conscious engineering principles to minimize resource consumption and environmental impact throughout the product lifecycle

Axiom™ SwiftArray™ Kits: Advanced Array Processing Solutions

Industry-Leading Performance with Cutting-Edge Array Content

The Axiom™ SwiftArray™ kits deliver best-in-class turnaround time while providing access to the most up-to-date array content organized by specific application requirements. This combination of rapid processing capabilities and current genomic content ensures that researchers can achieve timely results without compromising on the relevance or comprehensiveness of their array-based analyses. The optimized performance characteristics make these kits particularly valuable in time-sensitive research environments where both speed and data currency are critical factors.

Integrated SwiftArrayStudio Solution Technology

Built around the SwiftArrayStudio Solution technology platform, these kits represent a comprehensive approach to array processing that integrates hardware, software, and reagent components into a unified system. The SwiftArrayStudio Solution provides the technological foundation that enables the superior performance characteristics of the Axiom™ SwiftArray™ system, ensuring seamless integration between all components of the workflow. This integrated approach eliminates compatibility concerns and optimizes system performance across all processing steps.

Specialized Compatibility with Custom Axiom SwiftArrays

The Axiom™ SwiftArray™ kits are specifically engineered for use with custom Axiom SwiftArrays, ensuring optimal performance when paired with these specialized array formats. This targeted compatibility means that researchers utilizing custom array designs can achieve maximum efficiency and reliability in their sample processing workflows. The kits' formulation takes into account the unique requirements of custom array configurations, providing the precise reagent compositions needed to support these specialized applications.

Optimized Reagent Systems for High-Performance Processing

Each Axiom™ SwiftArray™ kit contains carefully optimized reagents and buffers specifically formulated for use with the AppliedBiosystems SwiftArrayStudio Instrument and Axiom™ SwiftArray™ assays. These reagent systems are designed to support efficient, high-performance array processing through streamlined workflows that consistently deliver reliable results. The optimization extends to both standard laboratory settings and higher throughput environments, ensuring that performance remains consistent regardless of processing volume requirements.

Flexible Configuration Options for Diverse Throughput Requirements

The kits are available in multiple format configurations, including 1x and 2x options, allowing laboratories to select the appropriate scale to match their specific throughput needs. Additionally, optional non-wash-inclusive versions are available to provide further flexibility for workflow management procedures. This flexibility in kit configurations enables laboratories to optimize their inventory management while ensuring that their chosen format aligns precisely with their operational requirements and processing volumes.

TRUSTED_SUSTAINABILITY

Specifications

Type Reagent Kit
Array Genotyping
Number of Arrays 1 array
Format 96-well Plate, Format 96
Species Human
No. of Samples 2x96F
Quantity 192 Reactions

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.