Learn More
Zeiss™ Axioscope 5 Upright Microscope
Product Code. 17362810
Change view
Click to view available options
For Use With (Application):
Transmitted Light Brightfield and DIC
Unit Size:
1 set
Product Code. For Use With (Application) unitSize
17362810 Transmitted Light Brightfield and DIC 1 set
1 options
Product Code. 17362810 Supplier Zeiss™ Supplier No. 4900400048000
 Restricted Item
This item requires a price update prior to being ordered.
Request a Price for this item here.
Flexible upright microscope ideal for biomedical routine & research. Configurable for the clinical environment. System is configured for transmitted brightfield & DIC microscopy with phototube. Use in combination with Axiocam 208 color for a smart standalone microscope system.
  • The Axioscope 5 is a flexible manual upright microscope that can be used for a for a range of transmitted light observations and also a fluorescence model is available. The possible contrast methods are brightfield, darkfield, phase contrast, PlasDIC, DIC, fluorescence,
    and transmitted/reflected light polarization depending on the configuration. This allows the microscope to be used for a range of applications in biology research, human and veterinary medicine, microbiology, plant science & botany and forensics.
  • The microscope body is powder coated durable metal, heat treated and resistant to standard reagents used for staining, organic solvents used for cleaning of lenses and disinfectants.
  • All optical parts including objectives, eyepieces, lenses, prisms, have anti-reflective and anti-fungal coating.
  • The microscope offers a coaxial z-drive with coarse and fine focus. Fine focus knob left and right.The focus range is 24 mm. The microscope includes an adjustable focus stop. The coarse focus provides movements of 4 mm/rotation, the fine focus comes with 400μm/rotation and with a graduation in micrometers (4μm).
  • The microscope comes with a manual revolving nosepiece with 6 encoded positions, suitable for reading objective position for light intensity manager functionality and for scaling information.
  • The microscope provides an energy saving and illumination life time extending Eco-mode function. When active, your microscope automatically goes to standby after being idle for 15 minutes, with a click of any button the microscope system is instantly on again.
  • The microscope body is equipped with two integrated, ergonomically positioned “snap image buttons” close to the focus drive on both sides of the microscope body to minimize hand movements and to optimally support left and right handed users. The snap image button allows to acquire images and videos directly from the stand (with Axiocam 208/202 in stand-alone mode and with Labscope; with any Axiocam with ZEN).
Specifications
For Use With (Application) Transmitted Light Brightfield and DIC