missing translation for 'onlineSavingsMsg'
Learn More

Axygen™ 10μL Microvolume Filter Tips

Product Code. 11324255 Shop All Axygen Scientific Products
Click to view available options
Volume (Metric):
0.5 to 10 μL
Unit Size:
Pack of 1000
This item is not returnable. View return policy

Product Code. 11324255

Brand: Axygen™ undefined

Only null left
Add to basket
Add to basket
This item is not returnable. View return policy

Axygen Universal Tips designed to fit a wide variety of single and multi-channel pipettes, compatible with most popular brands of pipettes. These polypropylene tips are nonpyrogenic and certified to be RNase-/DNase-free.

Axygen™ Universal Tips are designed to fit a wide variety of single and multi-channel pipettors, compatible with most popular brands of pipettors. These polypropylene tips are nonpyrogenic and certified to be RNase-/DNase-free. They are available in Maxymum Recovery, beveled, nonsterile, sterile, non-filtered, and other features for specialized applications. Flexible walls and a series of internal sealing rings ensure a secure fit with less force required to load and eject.

  • These polypropylene tips are nonpyrogenic and certified to be RNase-/DNase-free.
  • Nonpyrogenic
  • Microvolume
  • Maxymum Recovery technology for ultra-low retention and high yield
TRUSTED_SUSTAINABILITY

Specifications

Volume (Metric) 0.5 to 10 μL
Filtered Filtered
Sterility Non-sterile
Color Clear
Certifications/Compliance USP 3127
Clean Claims DNase, Rnase Free
For Use With (Application) Allows recovery of costly samples with improved precision and accuracy
Tip Style Low Retention
Packaging 1,000 Tips/Pk., 20 Packs/Cs.
For Use With (Equipment) Gilson Pipetman P2, Gilson Pipetman P10
Material Polypropylene
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.