missing translation for 'onlineSavingsMsg'
Learn More

Axygen™ 1000 μL Universal Pipetter Tips: Bevelled

Product Code. 12697215 Shop All Axygen Scientific Products
Change view
Click to view available options
Sterility:
Non-sterile
Sterile
Color:
Blue
Clear
Format:
Bulk
Rack
Unit Size:
Pack of 1000
Pack of 5000
9 product options available for selection
Product selection table with 9 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Sterility Color Format unitSize
12697215 Non-sterile Clear Rack Pack of 1000
12176870 Non-sterile Blue Bulk Pack of 1000
12551308 Non-sterile Blue Bulk Pack of 5000
12003989 Non-sterile Blue Rack Pack of 5000
10342010 Sterile Blue Rack Pack of 5000
12186870 Non-sterile Clear Bulk Pack of 1000
12561308 Non-sterile Clear Bulk Pack of 5000
12712936 Non-sterile Clear Rack Pack of 5000
12722936 Sterile Clear Rack Pack of 5000
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 9 options available.
9 options
This item is not returnable. View return policy
Product Code. 12697215 Supplier Axygen™ Supplier No. T1000CR
Only null left
Add to basket
Edge
Add to basket
This item is not returnable. View return policy

Compatible with most popular brands of pipettes

Axygen™ Universal Tips are designed to fit a wide variety of single and multi-channel pipettors, compatible with most popular brands of pipettors. These polypropylene tips are nonpyrogenic and certified to be RNase-/DNase-free. They are available in Maxymum Recovery, beveled, nonsterile, sterile, non-filtered, and other features for specialized applications. Flexible walls and a series of internal sealing rings ensure a secure fit with less force required to load and eject.

  • These polypropylene tips are nonpyrogenic and certified to be RNase-/DNase-free.
  • Nonpyrogenic
  • Beveled
TRUSTED_SUSTAINABILITY

Specifications

Volume (Metric) 100 to 1000 μL
Sterility Non-sterile
Color Clear
Autoclavable Autoclavable
Clean Claims DNase, RNase, and Pyrogen free
Format Rack
Tip Style Beveled, Universal
Packaging Rack
For Use With (Equipment) Universal Fit
Material Polypropylene
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.