missing translation for 'onlineSavingsMsg'
Learn More

Axygen™ 1000 μL Universal Pipetter Tips: Wide Bore
GREENER_CHOICE

Product Code. 12581308 Shop All Axygen Scientific Products
Click to view available options
Sterility:
Non-sterile
Sterile
Format:
Bulk
Rack
Unit Size:
Pack of 5000
This item is not returnable. View return policy

Product Code. 12581308

Brand: Axygen™ T1005WBC

Only null left
Add to basket
Add to basket
This item is not returnable. View return policy

Compatible with most popular brands of pipettes

Axygen™ Universal Tips are designed to fit a wide variety of single and multi-channel pipettors, compatible with most popular brands of pipettors. These polypropylene tips are nonpyrogenic and RNase-/DNase-free. They are available in Maxymum Recovery, beveled, nonsterile, sterile, non-filtered, and other features for specialized applications. Flexible walls and a series of internal sealing rings ensure a secure fit with less force required to load and eject.

(This is the suggested replacement for T-1005-WB-C-L)

  • These polypropylene tips are nonpyrogenic and RNase-/DNase-free.
  • Nonpyrogenic
  • Wide-Bore
TRUSTED_SUSTAINABILITY

Specifications

Volume (Metric) 100 to 1000 μL
Filtered Non-filtered
Sterility Non-sterile
Color Clear
Autoclavable Autoclavable
Clean Claims DNase, RNase, DNA, and PCR inhibitor Free
Format Bulk
Tip Style Genomic/Wide Orifice, Universal
Packaging 1,000 Tips/Pk., 5 Packs/Cs.
For Use With (Equipment) Universal Fit
Material Polypropylene
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.