missing translation for 'onlineSavingsMsg'
Learn More

Axygen™ 200μL Universal Pipette Tips: 200μL, Beveled
GREENER_CHOICE

Product Code. 11376144 Shop All Axygen Scientific Products
Change view
Click to view available options
Sterility:
Non-sterile
Sterile
Color:
Clear
Yellow
Format:
Bulk
Hinged Rack
Rack
Rack, 5 x 96
Stack Rack
Unit Size:
Pack of 1000
Pack of 20000
Pack of 4800
Pack of 960
9 product options available for selection
Product selection table with 9 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Sterility Color Format unitSize
11376144 Non-sterile Clear Bulk Pack of 1000
12591288 Non-sterile Clear Rack, 5 x 96 Pack of 4800
12753109 Non-sterile Clear Hinged Rack Pack of 4800
12669595 Non-sterile Clear Hinged Rack Pack of 960
12083859 Sterile Clear Hinged Rack Pack of 4800
12521298 Sterile Yellow Stack Rack Pack of 4800
12783109 Sterile Yellow Rack Pack of 4800
12571358 Non-sterile Yellow Bulk Pack of 20000
12126880 Non-sterile Yellow Bulk Pack of 1000
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 9 options available.
9 options
This item is not returnable. View return policy
Product Code. 11376144 Supplier Axygen™ Supplier No. T200C
Only null left
Add to basket
Edge
Add to basket
This item is not returnable. View return policy

Designed to fit a wide variety of single and multi-channel pipettes.

  • Optimized tip bevel reduces sample hang-up, improves pipetting accuracy
  • Wide bore tips of orifice size: 0.062 in. (1.52 mm) makes it ideal for pipetting viscous materials, genomic DNA or delicate cell cultures
  • Graduation marks allow a visual confirmation of pipettor draw volume
  • Presterilized tips are e-beam irradiated to prevent product degradation
  • Clean-room manufacturing environment certifies products safe from RNase, DNase and pyrogen safe
  • Also available with Maxymum recovery, which allows complete retrieval of precious samples, saves costly reagents

Order Info

Available in bulk, racked or stack rack formats.

TRUSTED_SUSTAINABILITY

Specifications

Volume (Metric) 1 to 200 μL
Sterility Non-sterile
Color Clear
Autoclavable Autoclavable
Clean Claims DNase, RNase, DNA, and PCR inhibitor Free
Format Bulk
Tip Style Universal, Beveled
For Use With (Equipment) Universal Fit
Material Polypropylene
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.