missing translation for 'onlineSavingsMsg'
Learn More

Axygen™ 200μL Universal Pipette Tips: 200μL, Beveled

Product Code. 12126880 Shop All Axygen Scientific Products
Change view
Click to view available options
Sterility:
Non-sterile
Sterile
Color:
Clear
Yellow
Format:
Bulk
Hinged Rack
Rack
Rack, 5 x 96
Stack Rack
Unit Size:
Pack of 1000
Pack of 20000
Pack of 4800
Pack of 960
10 product options available for selection
Product selection table with 10 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Sterility Color Format Unit Size
12126880 Non-sterile Yellow Bulk Pack of 1000
11376144 Non-sterile Clear Bulk Pack of 1000
12591288 Non-sterile Clear Rack, 5 x 96 Pack of 4800
12753109 Non-sterile Clear Hinged Rack Pack of 4800
12669595 Non-sterile Clear Hinged Rack Pack of 960
12687215 Sterile Clear Hinged Rack Pack of 960
12083859 Sterile Clear Hinged Rack Pack of 4800
12521298 Sterile Yellow Stack Rack Pack of 4800
12783109 Sterile Yellow Rack Pack of 4800
12571358 Non-sterile Yellow Bulk Pack of 20000
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 10 options available.
10 options
This item is not returnable. View return policy
You may only order up to the available stock. Where an alternative item has been identified it will be shown below
Product Code. 12126880 Supplier Axygen™ Supplier No. T200Y
  • ONLINE EXCLUSIVE PRICE
    32.15 €
    29.05 € / Pack of 1000
    Save 3.10 €
    9% Off
View all promotions
In Stock
Add to basket
Edge
Add to basket
Bulk Quote Request

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy
You may only order up to the available stock. Where an alternative item has been identified it will be shown below

Designed to fit a wide variety of single and multi-channel pipettes.

  • Optimized tip bevel reduces sample hang-up, improves pipetting accuracy
  • Wide bore tips of orifice size: 0.062 in. (1.52 mm) makes it ideal for pipetting viscous materials, genomic DNA or delicate cell cultures
  • Graduation marks allow a visual confirmation of pipettor draw volume
  • Presterilized tips are e-beam irradiated to prevent product degradation
  • Clean-room manufacturing environment certifies products safe from RNase, DNase and pyrogen safe
  • Also available with Maxymum recovery, which allows complete retrieval of precious samples, saves costly reagents

Order Info

Available in bulk, racked or stack rack formats.

TRUSTED_SUSTAINABILITY

Specifications

Volume (Metric) 1 to 200 μL
Filtered Non-filtered
Sterility Non-sterile
Color Yellow
Autoclavable Autoclavable
Model T-200-Y
Certifications/Compliance EN ISO 13485 compliant, BSE/TSE compliant, Non-Pyrogenic, USP Class VI tested, Human DNA Free, DNase/RNase Free, Quality Control Tested
Clean Claims DNase, RNase, and Pyrogen free
Format Bulk
Tip Style Beveled, Universal
Quantity 1000/Pk., 20 Pk./Cs.
Product Type Pipette Tips
For Use With (Equipment) Universal Fit
Graduated Yes
Material Polypropylene
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.