missing translation for 'onlineSavingsMsg'
Learn More

Axygen™ 200μL Universal Pipette Tips: 200μL, Beveled
GREENER_CHOICE

Product Code. p-4102794 Shop All Axygen Scientific Products
Click to view available options
Sterility:
Non-sterile
Sterile
Color:
Clear
Yellow
Format:
Bulk
Hinged Rack
Rack
Rack, 5 x 96
Unit Size:
Pack of 1000
Pack of 20000
Pack of 4800
Pack of 960
This item is not returnable. View return policy

Product Code. 11376144

Brand: Axygen™ T200C

Only null left
Add to basket
Add to basket
This item is not returnable. View return policy

Designed to fit a wide variety of single and multi-channel pipettes.

  • Optimized tip bevel reduces sample hang-up, improves pipetting accuracy
  • Wide bore tips of orifice size: 0.062 in. (1.52 mm) makes it ideal for pipetting viscous materials, genomic DNA or delicate cell cultures
  • Graduation marks allow a visual confirmation of pipettor draw volume
  • Presterilized tips are e-beam irradiated to prevent product degradation
  • Clean-room manufacturing environment certifies products safe from RNase, DNase and pyrogen safe
  • Also available with Maxymum recovery, which allows complete retrieval of precious samples, saves costly reagents

Order Info

Available in bulk, racked or stack rack formats.

TRUSTED_SUSTAINABILITY

Specifications

Volume (Metric) 1 to 200 μL
Sterility Non-sterile
Color Clear
Autoclavable Autoclavable
Clean Claims DNase, RNase, DNA, and PCR inhibitor Free
Format Bulk
Tip Style Universal, Beveled
For Use With (Equipment) Universal Fit
Material Polypropylene
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.