missing translation for 'onlineSavingsMsg'
Learn More

Axygen™ 200 μL Universal Pipetter Tips: 200 μL, Beveled, Reference
GREENER_CHOICE

Product Code. 12126890 Shop All Axygen Scientific Products
Click to view available options
Sterility:
Non-sterile
Sterile
Color:
Clear
Yellow
Format:
Bulk
Rack
Refill
Unit Size:
Pack of 1000
Pack of 20000
Pack of 4800
Pack of 960
This item is not returnable. View return policy

Product Code. 12126890

Brand: Axygen™ TR222C

Only null left
Add to basket
Edge
Add to basket
This item is not returnable. View return policy

Designed to fit a wide variety of single and multi-channel pipettes

Axygen™ Universal Tips are designed to fit a wide variety of single and multi-channel pipettors. These polypropylene tips are nonpyrogenic and certified to be RNase-/DNase-free. They are available in Maxymum Recovery, beveled, nonsterile, sterile, non-filtered, and other features for specialized applications. Flexible walls and a series of internal sealing rings ensure a secure fit with less force required to load and eject.

(This is the suggested replacement for T-205-WB-C-L)

  • These polypropylene tips are nonpyrogenic and certified to be RNase-/DNase-free.
  • Nonpyrogenic
  • Wide-Bore

Order Info

Available in bulk, racked or stack rack formats.

TRUSTED_SUSTAINABILITY

Specifications

Volume (Metric) 1 to 200 μL
Filtered Non-filtered
Sterility Non-sterile
Color Clear
Autoclavable Autoclavable
Clean Claims DNase, RNase, DNA, and PCR inhibitor Free
Format Bulk
Tip Style Beveled, Universal
Packaging 1,000 Tips/Pk., 20 Packs/Cs.
For Use With (Equipment) Universal Fit
Graduated Yes
Material Polypropylene
Graduations 10, 50, 100μL
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.