missing translation for 'onlineSavingsMsg'
Learn More

Axygen™ 8-Strip PCR Tubes, 0.2 mL

Product Code. 12681646 Shop All Axygen Scientific Products
Change view
Click to view available options
Color:
Assorted
Blue
Clear
Green
Orange
Red
Violet
Yellow
Closure Type:
Domed Cap
Flat Cap
Sold Separately
Quantity:
100/Cs.
125/Pk., 1250/Cs.
Unit Size:
Pack of 100
Pack of 125
Pack of 1250
14 product options available for selection
Product selection table with 14 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Color Closure Type Quantity Unit Size
12681646 Clear Sold Separately 125/Pk., 1250/Cs. Pack of 125
12706839 Blue Sold Separately 125/Pk., 1250/Cs. Pack of 1250
10841291 Clear Sold Separately 125/Pk., 1250/Cs. Pack of 1250
12667215 Clear Domed Cap 125/Pk., 1250/Cs. Pack of 125
10810721 Clear Domed Cap 125/Pk., 1250/Cs. Pack of 1250
12796829 Assorted Sold Separately 125/Pk., 1250/Cs. Pack of 1250
12657215 Clear Flat Cap 125/Pk., 1250/Cs. Pack of 125
12756839 Green Sold Separately 125/Pk., 1250/Cs. Pack of 1250
12587977 Green Sold Separately 125/Pk., 1250/Cs. Pack of 125
12766839 Orange Sold Separately 125/Pk., 1250/Cs. Pack of 1250
12786839 Violet Sold Separately 125/Pk., 1250/Cs. Pack of 1250
12796839 Yellow Sold Separately 125/Pk., 1250/Cs. Pack of 1250
12776839 Red Sold Separately 125/Pk., 1250/Cs. Pack of 1250
15479063 Clear Sold Separately 100/Cs. Pack of 100
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 14 options available.
14 options
This item is not returnable. View return policy
Product Code. 12681646 Supplier Axygen™ Supplier No. PCR0208C
  • ONLINE EXCLUSIVE PRICE
    140.00 €
    135.28 € / Pack of 125
    Save 4.72 €
    3% Off
In Stock
Add to basket
Edge
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Available in a variety of colors for easy sample identification. Axygen™ 8-Strip PCR Tubes, 0.2mL are constructed of sturdy polypropylene. Each non-sterile, conical tube features thin and consistent wall thickness. Sold with flat or domed caps to prevent sample loss or without caps.

Available in a variety of colors for easy sample identification. Axygen™ 8-Strip PCR Tubes, 0.2mL are constructed of sturdy polypropylene. Each non-sterile, conical tube features thin and consistent wall thickness. Sold with flat or domed caps to pr

  • PCR tubes strips designed to fit a wide variety of thermal cyclers
  • Ultra-thin and consistent wall thickness allows precise thermal transfer
  • Domed or flat insertion caps ensure a tight fit that prevents sample evaporation
  • Certified RNase-/DNase-free and nonpyrogenic
  • Manufactured with 99.9% pure virgin polypropylene
  • Available in a variety of colors for easy sample identification
TRUSTED_SUSTAINABILITY

Specifications

Certifications/Compliance RNase, DNase, pyrogen-free
Color Clear
Volume (Metric) 0.2 mL
Quantity 125/Pk., 1250/Cs.
Closure Material Polypropylene
For Use With (Equipment) Thermal cyclers for thermal transfer applications
Shape Conical
Sterility Non-sterile
Material Polypropylene
Max. Force 4000 x g
Closure Type Sold Separately
Format Strip
Product Type Thin Wall 8-Strip PCR Tube
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.