missing translation for 'onlineSavingsMsg'
Learn More

Axygen™ Biomek™ FX/NX Robotic Tips
BIOPHARMA

Product Code. 12742856 Shop All Axygen Scientific Products
Change view
Click to view available options
Volume (Metric):
10 μL
20 μL
30 μL
45 μL
50 μL
250 μL
Sterility:
Non-sterile
Sterile
Tip Style:
Automation, Fine Point
Automation, Round
Automation, Wide Orifice
Format:
Rack, 384-well
Rack, 96
Unit Size:
Pack of 19200
Pack of 4800
Pack of 960
24 product options available for selection
Product selection table with 24 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Volume (Metric) Sterility Tip Style Format unitSize
12742856 250 μL Sterile Automation, Round Rack, 96 Pack of 4800
12732846 10 μL Non-sterile Automation, Round Rack, 96 Pack of 4800
12742846 10 μL Sterile Automation, Round Rack, 96 Pack of 4800
12792846 20 μL Non-sterile Automation, Round Rack, 96 Pack of 4800
12702856 20 μL Sterile Automation, Round Rack, 96 Pack of 4800
12772846 20 μL Non-sterile Automation, Round Rack, 96 Pack of 4800
12599167 20 μL Non-sterile Automation, Round Rack, 96 Pack of 960
12782846 20 μL Sterile Automation, Round Rack, 96 Pack of 4800
15591383 30 μL Non-sterile Automation, Fine Point Rack, 384-well Pack of 19200
12722866 30 μL Sterile Automation, Round Rack, 384-well Pack of 19200
12712866 30 μL Non-sterile Automation, Round Rack, 384-well Pack of 19200
15501393 30 μL Sterile Automation, Fine Point Rack, 384-well Pack of 19200
12792856 30 μL Non-sterile Automation, Round Rack, 384-well Pack of 19200
12782856 45 μL Sterile Automation, Fine Point Rack, 384-well Pack of 19200
12772856 45 μL Non-sterile Automation, Fine Point Rack, 384-well Pack of 19200
12772866 50 μL Non-sterile Automation, Round Rack, 96 Pack of 4800
12782866 50 μL Sterile Automation, Round Rack, 96 Pack of 4800
12792866 50 μL Non-sterile Automation, Round Rack, 96 Pack of 4800
12702876 50 μL Sterile Automation, Round Rack, 96 Pack of 4800
12712856 250 μL Non-sterile Automation, Round Rack, 96 Pack of 4800
12722856 250 μL Sterile Automation, Round Rack, 96 Pack of 4800
12732856 250 μL Non-sterile Automation, Round Rack, 96 Pack of 4800
12752856 250 μL Non-sterile Automation, Wide Orifice Rack, 96 Pack of 4800
12762856 250 μL Sterile Automation, Wide Orifice Rack, 96 Pack of 4800
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 24 options available.
24 options
This item is not returnable. View return policy
Product Code. 12742856 Supplier Axygen™ Supplier No. FX250RS
Add to basket
Add to basket
This item is not returnable. View return policy

Axygen robotic tips are manufactured to stringent specifications under strict process controls and assembled via automation to ensure consistent performance and quality.

The Axygen brand range of products includes a comprehensive line of tips for Beckman Coulter automated liquid handling systems. These tips are manufactured to stringent specifications under strict process controls and assembled via automation to ensure consistent performance and quality. The pipette tips are also available with MAXYMum Recovery, a unique low-retention technology that improves tip performance and maximizes the use of your precious samples.

  • Available in racked, racked sterile and with aerosol barrier (filtered)
  • Compatible with Beckman Biomek FX and Beckman Multimek instruments with an AP96 head
  • Also compatible with Span-8 pod when Liquid Level Sensing function is turned off
  • Unsurpassed straightness to ensure tip enters the well smoothly, every time
  • 384/1536-well compatible to meet your most demanding applications
  • Wide bore tips are ideal for pipetting viscous materials, genomic DNA or delicate cell cultures
  • Fine Point improves accuracy and precision at lower sample volumes
  • Maxymum recovery™ tips reduce sample loss, improves reproducibility and accuracy, minimizes usage of precious samples and reagents

TRUSTED_SUSTAINABILITY

Specifications

Volume (Metric) 250 μL
Filtered Non-filtered
Sterility Sterile
Color Clear
Autoclavable Not Autoclavable
Certifications/Compliance ANSI
Clean Claims DNase, RNase and Pyrogen free
For Use With (Application) Non-pyrogenic, used for stringent specifications under strict process controls
Format Rack, 96
Tip Style Automation, Round
Packaging 10 Racks/Pk, 5 Pks/Cs.
For Use With (Equipment) Beckman Coulter Biomek FX, Beckman Coulter Biomek NX, Beckman Coulter Biomek 3000, Beckman Coulter Multimek AP96, Beckman Coulter Multimek SPAN-8, Beckman Coulter Multimek MP20, Beckman Coulter Multimek MP200
Product Line Beckman Biomek
Material Polypropylene
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.