missing translation for 'onlineSavingsMsg'
Learn More

Axygen™ Disposable V-Bottom Reagent Reservoirs
GREENER_CHOICE

Product Code. 12507977 Shop All Axygen Scientific Products
Click to view available options
Capacity (Metric):
25 mL
50 mL
100 mL
Sterility:
Non-sterile
Sterile
Packaging:
100/Cs.
200/Cs.
Divided; 100/Cs.
Divided; 200/Cs.
Divided; Individually Wrapped; 100/Cs.
Individually Wrapped; 100/Cs.
Unit Size:
Pack of 100
Pack of 200
This item is not returnable. View return policy

Product Code. 12507977

Brand: Axygen™ RESV50SI

Only null left
Add to basket
Add to basket
This item is not returnable. View return policy

Designed to accommodate all multichannel pipettor

Axygen disposable reservoirs are designed to accommodate all multichannel pipettors. Four corners and a sharp V-bottom shape ensure maximum sample retrieval. All reservoirs are white.

TRUSTED_SUSTAINABILITY

Specifications

Certifications/Compliance ANSI, RNase/DNase-free, non-pyrogenic
Color White
Capacity (Metric) 50 mL
Material Polystyrene
Sterility Sterile
Type Reagent Reservoir
No. per Case 100/Cs.
No. per Pack 100/Pk.
Packaging Individually Wrapped; 100/Cs.
Disposable Single-use
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.