missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Axygen™ Disposable V-Bottom Reagent Reservoirs
Shop All Axygen Scientific Products

Click to view available options
Capacity (Metric):
25 mL
50 mL
100 mL
Sterility:
Non-sterile
Sterile
Packaging:
100/Cs.
200/Cs.
Divided; 100/Cs.
Divided; 200/Cs.
Divided; Individually Wrapped; 100/Cs.
Individually Wrapped; 100/Cs.
Unit Size:
Pack of 100
Pack of 200
Description
Designed to accommodate all multichannel pipettor
Axygen disposable reservoirs are designed to accommodate all multichannel pipettors. Four corners and a sharp V-bottom shape ensure maximum sample retrieval. All reservoirs are white.
Specifications
Specifications
| Certifications/Compliance | ANSI, RNase/DNase-free, non-pyrogenic |
| Color | White |
| Capacity (Metric) | 100 mL |
| Material | Polystyrene |
| Sterility | Sterile |
| Type | Reagent Reservoir |
| No. per Case | 100/Cs. |
| No. per Pack | 100/Pk. |
| Packaging | Individually Wrapped; 100/Cs. |
| Disposable | Single-use |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction