missing translation for 'onlineSavingsMsg'
Learn More

Axygen™ Disposable V-Bottom Reagent Reservoirs

Product Code. 12567997 Shop All Axygen Scientific Products
Change view
Click to view available options
Capacity (Metric):
25 mL
50 mL
100 mL
Sterility:
Non-sterile
Sterile
Packaging:
100/Cs.
200/Cs.
Divided; 100/Cs.
Divided; 200/Cs.
Divided; Individually Wrapped; 100/Cs.
Individually Wrapped; 100/Cs.
Unit Size:
Pack of 100
Pack of 200
12 product options available for selection
Product selection table with 12 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Capacity (Metric) Sterility Packaging Unit Size
12567997 50 mL Sterile 200/Cs. Pack of 200
12537967 25 mL Non-sterile Divided; 100/Cs. Pack of 100
12537997 25 mL Sterile Divided; 200/Cs. Pack of 200
12547967 25 mL Sterile Divided; Individually Wrapped; 100/Cs. Pack of 100
12547997 100 mL Sterile 200/Cs. Pack of 200
12567967 100 mL Sterile Individually Wrapped; 100/Cs. Pack of 100
12557967 100 mL Non-sterile 100/Cs. Pack of 100
10241570 25 mL Non-sterile 100/Cs. Pack of 100
12557997 25 mL Sterile 200/Cs. Pack of 200
12587967 25 mL Sterile Individually Wrapped; 100/Cs. Pack of 100
12597967 50 mL Non-sterile 100/Cs. Pack of 100
12507977 50 mL Sterile Individually Wrapped; 100/Cs. Pack of 100
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 12 options available.
12 options
This item is not returnable. View return policy
Product Code. 12567997 Supplier Axygen™ Supplier No. RESV50S
  • ONLINE EXCLUSIVE PRICE
    207.81 €
    188.90 € / Pack of 200
    Save 18.91 €
    9% Off
View all promotions
In Stock
Add to basket
Edge
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Designed to accommodate all multichannel pipettor

Axygen disposable reservoirs are designed to accommodate all multichannel pipettors. Four corners and a sharp V-bottom shape ensure maximum sample retrieval. All reservoirs are white.

TRUSTED_SUSTAINABILITY

Specifications

Certifications/Compliance ANSI, RNase/DNase-free, non-pyrogenic
Color White
Capacity (Metric) 50 mL
Material Polystyrene
Sterility Sterile
Type Reagent Reservoir
Packaging 200/Cs.
Disposable Single-use
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.