missing translation for 'onlineSavingsMsg'
Learn More

Axygen™ IMAG Handheld Magnetic Beads Separation Devices

Product Code. 15879692 Shop All Corning Products
Change view
Click to view available options
Format:
96-well Plate
Tube
Unit Size:
Each
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Format Unit Size
15879692 96-well Plate Each
17246740 Tube Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15879692 Supplier Axygen™ Supplier No. IMAG96P
  • ONLINE EXCLUSIVE PRICE
    1405.00 €
    1365.47 € / Pack of 1
    Save 39.53 €
    2% Off
Estimated Shipment: 30-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Transform low throughput to high throughput separation by a flip of a tube or microplate

The use of magnetic separation devices (MSDs) are essential in any paramagnetic beads-based purification process. Traditionally, MSDs are not optimized for manual use and most require electrically powered liquid handling systems. IMAG MSDs are available in tube or microplate format and are designed to simplify the manual processing of magnetic bead separations, including nucleic acid purification and clean-up, cell-based assays, and antibody and protein purifications.

  • IMAG MSDs accommodate single tube or 96-well microplate formats
  • Fast separation time

        - Strong magnets enable less than 30-second separation times

        - Holds tubes or microplate enabling fast wash steps

  • Manual operation enables low or high throughput
TRUSTED_SUSTAINABILITY

Specifications

Format 96-well Plate
Isolation Technology Magnetic Bead
For Use With (Equipment) 96 Microplates
For Use With (Application) Nucleic Acid purification and clean up, cell based assays, Antibody and Protein purification
Includes Strong magnets providing less than 30 sec. separation time
Product Type Magnetic Separation Device
Quantity Each
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.