missing translation for 'onlineSavingsMsg'
Learn More

Axygen™ Oasis™ Robotic Tips

Product Code. p-4102787 Shop All Axygen Scientific Products
Click to view available options
Volume (Metric):
100 μL
Rack Color:
Gray
Filtered:
Non-filtered
Unit Size:
Pack of 4800
This item is not returnable. View return policy

Product Code. 12772886

Brand: Axygen™ OT100R

Only null left
Add to basket
Add to basket
This item is not returnable. View return policy

For use with Oasis automated liquid handling systems

  • Tip format: 96 tip
  • Tip volume range: 50 μL to 200 μL
  • Tip material: Clear polypropylene
  • RNase-/DNase-/human gDNA-free
  • Nonpyrogenic
  • Available options:
    - With or without aerosol-resistant filters
    - Nonsterile or sterile
  • Compatible heads: 8- or 96-channel standard volume
  • Rack format: ANSI/SLAS
  • Rack material: Polypropylene
TRUSTED_SUSTAINABILITY

Specifications

Volume (Metric) 100 μL
Filtered Non-filtered
Sterility Non-sterile
Color Clear
Rack Color Gray
Clean Claims DNase, RNase Free
Format Rack
Tip Style Automation, Round
Packaging SBS Rack
Quantity 50 Racks/Cs.
For Use With (Equipment) Dynamic Devices Oasis
Product Line Oasis
Material Polyethylene
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.