missing translation for 'onlineSavingsMsg'
Learn More

Axygen™ PCR Tubes with 0.2 mL Dome Cap
GREENER_CHOICE

Product Code. 12756849 Shop All Axygen Scientific Products
Click to view available options
Color:
Assorted
Blue
Clear
Green
Orange
Red
Spectrum
Violet
White
Yellow
Product Type:
Maxymum Recovery, Domed Cap PCR Tubes
Thin Wall, Domed Cap PCR Tube
Unit Size:
Pack of 1000
Pack of 10000
This item is not returnable. View return policy

Product Code. 12756849

Brand: Axygen™ PCR02DA

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Thin and consistent wall thickness for precise thermal transfer

Axygen™ 0.2 mL thin-wall 12 strip PCR tubes, available in an assortment of colors are designed to fit a wide variety of thermal cyclers. Ultra-thin and consistent wall thickness allows precise thermal transfer, even insertion cap ensures a tight fit to prevent evaporation.


  • RNase-/DNase-free
  • Nonpyrogenic
  • Nonsterile
  • Caps sold separately
  • Manufactured with 99.9% pure virgin polypropylene
  • TRUSTED_SUSTAINABILITY

    Specifications

    Certifications/Compliance RNase, DNase, pyrogen-free
    Color Assorted
    Volume (Metric) 0.2 mL
    Quantity 1000/Pk., 10Pk./Cs.
    Closure Material Polypropylene
    For Use With (Equipment) Standard 96-well blocks of real-time PCR instruments and thermal transfer applications
    Shape Conical
    Sterility Non-sterile
    Material Polypropylene
    Closure Type Domed Cap
    Format Tube
    Product Type Thin Wall, Domed Cap PCR Tube
    Show More Show Less
    Product Content Correction

    Your input is important to us. Please complete this form to provide feedback related to the content on this product.

    Product Title

    By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

    Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.