missing translation for 'onlineSavingsMsg'
Learn More

Axygen™ Universal Fit Micropipette Tips

Product Code. 12726729 Shop All Axygen Scientific Products
Click to view available options
Sterility:
Non-sterile
Sterile
Color:
Clear
Yellow
Tip Style:
Beveled
Genomic/Wide Orifice
Low Retention, Beveled
Low Retention, Universal
Universal
Quantity:
10,000/Cs.
4800/Cs.
Unit Size:
Pack of 10000
Pack of 4800
This item is not returnable. View return policy

Product Code. 12726729

Brand: Axygen™ TE204YRS

Only null left
Add to basket
Edge
Add to basket
This item is not returnable. View return policy

Axygen™ Universal Fit Pipette Tips are compatible with most research-grade single- and multi-channel micropipettes, and are certified to be DNase-/RNase-free and nonpyrogenic.

  • RNase-/DNase free
  • Nonpyrogenic
  • Human gDNA- and PCR-inhibition-free
  • Available in a variety of sizes
  • Available with a variety of tip styles including beveled, fine point, and wide bore
  • Available sterile/non-sterile
  • Available in bulk, racked, or stack rack packaging
  • Available with Maxymum Recovery™ surface for improved accuracy and maximum sample recovery

Order Info

dimQuantity

TRUSTED_SUSTAINABILITY

Specifications

Volume (Metric) 200 μL
Filtered Non-filtered
Sterility Sterile
Color Yellow
Autoclavable Autoclavable
Clean Claims Dnase-, Rnase-, Pyrogen-, Human gDNA-, PCR-Inhibition-Free
Format Rack
Tip Style Universal
Packaging 96 x 10 Racks/Pk., 5 Pk./Cs.
Quantity 4800/Cs.
For Use With (Equipment) Universal Fit, Universal Fit, Eppendorf Compatible
Graduated Yes
Material Polypropylene
Graduations 10, 50, 100μL
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.