missing translation for 'onlineSavingsMsg'
Learn More

Axygen™ Aerosol Barrier Micropipette Tips

Product Code. 12726739 Shop All Axygen Scientific Products
Click to view available options
Volume (Metric):
0.5 to 50 μL
1 to 150 μL
100 to 1000 μL
Sterility:
Non-sterile
Sterile
Unit Size:
Pack of 1000
Pack of 10000
Pack of 4800
This item is not returnable. View return policy

Product Code. 12726739

Brand: Axygen™ TF150RS

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Axygen™ Aerosol Barrier Micropipette Tips allow free passage of air while blocking aerosol contamination.

  • Technologically advanced filter material
  • Filter membrane contains no leachable additives
  • Available with Maxymum Recovery™ surface for improved accuracy and maximum sample recovery
  • Available in bulk, or racked (sterile)
  • Available in a variety of material contact tip options
TRUSTED_SUSTAINABILITY

Specifications

Volume (Metric) 1 to 150 μL
Filtered Filtered
Sterility Sterile
Color Clear
Autoclavable Autoclavable
Clean Claims DNase, RNase Free
Format Rack
Tip Style Universal
Packaging 96 tips/rack, 10 racks/pack, 5 packs/case
Product Type Aerosol Barrier Micropipette Tips
For Use With (Equipment) Universal Fit
Material Polypropylene
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.