missing translation for 'onlineSavingsMsg'
Learn More

Axygen™ Screw Cap Tubes without Caps: Conical

Conical tubes

Brand:  Axygen™ ST-45-S

Code : 66

Additional Details : Weight : 6.80000kg

 View more versions of this product

Product Code. 12759618

  • 620.00 € / Pack of 4000
Estimated Shipment: 31-07-2024
to see stock.
Add to basket

Description

Description

Serve a broad range of laboratory needs including collection, processing, centrifugation, storage and packaging/shipping

  • Tube volumes from 0.5 to 2.0mL
  • Choose clear or amber

Specifications

Specifications

20,000 x g
0.5 mL
Polypropylene
Sterile
Conical
RNase, DNase, pyrogen-free
Clear
Bag
Not Included
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers