missing translation for 'onlineSavingsMsg'
Learn More

Axygen™ Screw Caps with O-rings

Product Code. 10692261 Shop All Axygen Scientific Products
Change view
Click to view available options
Color:
Blue
Brown
Clear
Gray
Green
Orange
Pink
Red
White
Yellow
Sterility:
Non-sterile
Sterile
Format:
Looped
Non-looped
Unit Size:
Pack of 4000
Pack of 500
25 product options available for selection
Product selection table with 25 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Color Sterility Format Unit Size
10692261 Yellow Non-sterile Non-looped Pack of 4000
12759368 Brown Non-sterile Non-looped Pack of 4000
11310325 Blue Non-sterile Non-looped Pack of 500
12769368 Brown Sterile Non-looped Pack of 4000
12799368 Clear Sterile Non-looped Pack of 4000
10328609 Green Non-sterile Non-looped Pack of 4000
12789368 Clear Non-sterile Non-looped Pack of 4000
12719378 Gray Non-sterile Non-looped Pack of 4000
12729378 Gray Sterile Non-looped Pack of 4000
12769378 Brown Non-sterile Looped Pack of 4000
12729388 Orange Non-sterile Looped Pack of 4000
12799378 Green Non-sterile Looped Pack of 4000
12759378 Blue Non-sterile Looped Pack of 4000
12779378 Clear Non-sterile Looped Pack of 4000
12709388 Gray Non-sterile Looped Pack of 4000
12739388 Pink Non-sterile Looped Pack of 4000
12789388 Yellow Non-sterile Looped Pack of 4000
12709398 Orange Sterile Non-looped Pack of 4000
12799388 Orange Non-sterile Non-looped Pack of 4000
12779388 White Non-sterile Looped Pack of 4000
12719398 Pink Non-sterile Non-looped Pack of 4000
12729398 Pink Sterile Non-looped Pack of 4000
10387849 Red Non-sterile Non-looped Pack of 4000
12789398 White Sterile Non-looped Pack of 4000
12709408 Yellow Sterile Non-looped Pack of 4000
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 25 options available.
25 options
This item is not returnable. View return policy
Product Code. 10692261 Supplier Axygen™ Supplier No. SCOY
  • ONLINE EXCLUSIVE PRICE
    830.00 €
    815.19 € / Pack of 4000
    Save 14.81 €
    1% Off
Estimated Shipment: 09-04-2027
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Select looped or non-looped format

Axygen microcentrifuge tubes feature a screw cap that provides a tight secure seal. Screw cap tubes are available in a variety of colors with a conical or self-standing bottom. Sterile options are also available.

  • Maximum G-force is 20,000 RCF.
  • RNase-/DNase-free
  • Nonpyrogenic


Tubes, with Cap, Sterile


All sterile screw cap microcentrifuge tubes are pre-assembled with non-looped caps with O-Rings.


Colored-cap versions are also available in sterile format.
TRUSTED_SUSTAINABILITY

Specifications

Includes O-ring
Product Type Screw Cap with O-ring
Material Polypropylene
Color Yellow
Format Non-looped
No. per Pack 500/Pk.
No. per Case 4000/Cs.
Sterility Non-sterile
For Use With (Equipment) Both skirted and non-skirted variations of the 0.5 mL, 1.5 mL, and 2 mL microcentrifuge tubes
Disposable Single-use
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.