missing translation for 'onlineSavingsMsg'
Learn More

Axygen™ Tecan™ Evo MCA96 Compatible Nested Pipet Tips
BIOPHARMA

Product Code. 15388291 Shop All Axygen Scientific Products
Change view
Click to view available options
Volume (Metric):
50 μL
200 μL
Sterility:
Non-sterile
Sterile
Color:
Gray
Orange
Unit Size:
Pack of 3840
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Volume (Metric) Sterility Color Unit Size
15388291 200 μL Sterile Gray Pack of 3840
15378291 200 μL Non-sterile Gray Pack of 3840
15277668 50 μL Sterile Orange Pack of 3840
15398291 50 μL Non-sterile Orange Pack of 3840
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 15388291 Supplier Axygen™ Supplier No. EV200NTRS
  • ONLINE EXCLUSIVE PRICE
    398.00 €
    328.11 € / Pack of 3840
    Save 69.89 €
    17% Off
Estimated Shipment: 09-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

768 tips for use with Tecan Evo MCA96 multi-channel arm

  • Tip format: 96 tip
  • Tip volume range: 50 μL to 200 μL
  • Tip material: Clear polypropylene
  • RNase-/DNase-/human gDNA-free
  • Nonpyrogenic
  • Available options:
    - With or without aerosol-resistant filters
    - Nonsterile or sterile
  • Compatible heads: MCA96
  • Rack format: ANSI/SLAS
  • Rack material: Polypropylene
TRUSTED_SUSTAINABILITY

Specifications

Volume (Metric) 200 μL
Sterility Sterile
Color Gray
Tip Style Round
Packaging Nested tip rack
Quantity 40 Racks/Cs.
For Use With (Equipment) Tecan EVO MCA96
Product Line Tecan
Material Polypropylene
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.