Learn More
Axygen™ Tip Refill System
Product Code. 12551288 Shop All Axygen Scientific Products
Change view
Click to view available options
Volume (Metric):
0.1 to 10 μL
1 to 200 μL
15 to 1250 μL
Unit Size:
Pack of 4608
Pack of 4800
Pack of 768
Pack of 960
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Volume (Metric) unitSize
12551288 1 to 200 μL Pack of 4800
12511288 15 to 1250 μL Pack of 4608
11300285 15 to 1250 μL Pack of 768
11310285 1 to 200 μL Pack of 960
12561288 0.1 to 10 μL Pack of 4800
11320285 0.1 to 10 μL Pack of 960
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Product Code. 12551288 Supplier Axygen™ Supplier No. RFL222C
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Axygen™ style and universal fit pipet tips in easy-to-load refill racks
Axygen™ Tip Refill System contains durable refill rack that can be easily refilled by hand and is autoclavable. These translucent Racks includes color-coded inserts for tip identification and are certified RNase-/DNase free. They are available in different colors and are packaged in bulk or racks.
  • Certified to be RNase-/DNase-free
  • Nonpyrogenic
  • Translucent rack allows tips to be easily seen
  • Color-coded insert allows for easy identification of tip
  • TRUSTED_SUSTAINABILITY
    Specifications
    Volume (Metric) 1 to 200 μL
    Sterility Non-sterile
    Color Clear
    Autoclavable Autoclavable
    Tip Style Universal
    Packaging 96 Tips/Rack, 10 Racks/Pk, 5 Packs/Cs
    Graduated Yes