missing translation for 'onlineSavingsMsg'
Learn More

Axygen™ Tip Refill System

Product Code. 12561288 Shop All Axygen Scientific Products
Change view
Click to view available options
Volume (Metric):
0.1 to 10 μL
1 to 200 μL
15 to 1250 μL
Unit Size:
Pack of 4608
Pack of 4800
Pack of 768
Pack of 960
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Volume (Metric) unitSize
12561288 0.1 to 10 μL Pack of 4800
12511288 15 to 1250 μL Pack of 4608
11300285 15 to 1250 μL Pack of 768
11310285 1 to 200 μL Pack of 960
12551288 1 to 200 μL Pack of 4800
11320285 0.1 to 10 μL Pack of 960
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
This item is not returnable. View return policy
Product Code. 12561288 Supplier Axygen™ Supplier No. RFL300C

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Axygen™ style and universal fit pipet tips in easy-to-load refill racks

Axygen™ Tip Refill System contains durable refill rack that can be easily refilled by hand and is autoclavable. These translucent Racks includes color-coded inserts for tip identification and are certified RNase-/DNase free. They are available in different colors and are packaged in bulk or racks.
  • Certified to be RNase-/DNase-free
  • Nonpyrogenic
  • Translucent rack allows tips to be easily seen
  • Color-coded insert allows for easy identification of tip
  • TRUSTED_SUSTAINABILITY

    Specifications

    Volume (Metric) 0.1 to 10 μL
    Filtered Non-filtered
    Sterility Non-sterile
    Color Clear
    Autoclavable Autoclavable
    Model RFL-300-C
    Clean Claims RNase/DNase-free, Nonpyrogenic
    For Use With (Application) Diagnostics
    Packaging 96 Tips/Rack, 10 Racks/Pk, 5 Packs/Cs
    Quantity 96 Tips/Rack, 10 Racks/Pk., 5 Pk./Cs.
    Product Type Tip Refill System
    Includes Refill Rack
    For Use With (Equipment) Gilson Pipetman P10, Gilson Pipetman P2, Universal Fit
    Graduated No
    Material Polypropylene
    Show More Show Less
    Product Title
    Select an issue

    By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.