missing translation for 'onlineSavingsMsg'
Learn More

Axygen™ Automation Tips for Zymark™/Caliper, 25 μL
GREENER_CHOICE

Product Code. 12776679 Shop All Axygen Scientific Products
Click to view available options
Sterility:
Non-sterile
Unit Size:
Pack of 19200
This item is not returnable. View return policy

Product Code. 12776679

Brand: Axygen™ ZT384CR

Only null left
Add to basket
Add to basket
This item is not returnable. View return policy

Compatible with automated liquid handling systems. Axygen™ Automation Tips for Zymark™/Caliper, 25μL are non-filtered pipette tips in 384-well racks. Constructed of clear polypropyle under strict controls to ensure quality. Options include sterile and non-sterile tips.

  • Tip format: 96 tip and 384 tip
  • Tip volume range: 25 μL to 200 μL
  • Tip material: Clear polypropylene
  • RNase-/DNase-/human gDNA-free
  • Nonpyrogenic
  • Available options:
    - With or without aerosol-resistant filters
    - Nonsterile or sterile
    - Standard or Axygen™ Maxymum Recovery™ surface
  • Compatible heads: HV96, CAS-4200-1 (20 - 150 μL/sec.), LV96, LV384
  • Rack format: Non-ANSI (96) and ANSI/SLAS (384)
  • Rack material: Polypropylene
TRUSTED_SUSTAINABILITY

Specifications

Volume (Metric) 30 μL
Filtered Non-filtered
Sterility Non-sterile
Color Clear
Rack Color Light Green
Autoclavable Autoclavable
Clean Claims DNase, RNase Free
For Use With (Application) Non-pyrogenic
Format Rack
Tip Style Automation, Round
Packaging Non-SBS Rack
Quantity 50 Racks/Cs.
For Use With (Equipment) Zymark/Caliper RapidPlate 96/384, SciClone, Zephyr LV96, LV384 Head
Material Polypropylene
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.