missing translation for 'onlineSavingsMsg'
Learn More

Tocris Bioscience™ AY 77

Product Code. 18709956
Click to view available options
Quantity:
1 mg
Unit Size:
1mg
This item is not returnable. View return policy

Product Code. 18709956

Brand: Tocris Bioscience™ 7033/1

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Potent PAR2 biased agonist; exhibits bias for Ca2+ signaling

AY 77 is a potent PAR2 biased agonist. AY 77 exhibits bias for Ca2+ over ERK1/2 signaling (EC50 values are 40 nM for Ca2+ release versus 2 μM for ERK1/2 phosphorylation). Shows selectivity for PAR2 over PAR1. Promotes migration of human breast cancer cells in vitro.
TRUSTED_SUSTAINABILITY

Specifications

Purity 95%
CAS 1835734-92-3
Content And Storage Store at -20°C
Molecular Weight (g/mol) M.W.-observed: 404.5
Quantity 1 mg
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.