missing translation for 'onlineSavingsMsg'
Learn More

Tocris Bioscience™ AY 9944 dihydrochloride

Product Code. 18742704
Change view
Click to view available options
Quantity:
10 mg
Unit Size:
10mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
18742704 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18742704 Supplier Tocris Bioscience™ Supplier No. 1639/10
  • ONLINE EXCLUSIVE PRICE
    274.00 €
    258.30 € / 10mg
    Save 15.70 €
    5% Off
 Restricted Item
Estimated Shipment: 09-10-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Inhibitor of Hedgehog (Hh) signaling. Inhibits Δ7-dehydrocholesterol reductase

Inhibitor of hedgehog (Hh) signaling, possibly via several mechanisms. Inhibits Δ7-dehydrocholesterol reductase (IC50 = 13 nM), thus reduces cholesterol biosynthesis, and also inhibits cholesterol esterification. May also directly block the cellular response to Hh proteins. Teratogenic in vivo.
TRUSTED_SUSTAINABILITY

Specifications

CAS 366-93-8
Quantity 10 mg
Molecular Formula C22H30Cl4N2
Formula Weight 464.3
Purity >99%
Molecular Weight (g/mol) 464.296
Chemical Name or Material AY 9944 dihydrochloride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.